missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIP60 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534548
This item is not returnable.
View return policy
Description
A suggested positive control is human brain tissue lysate. PA5-34548 can be used with blocking peptide PEP-1591.
TIP60 is a transcription factor, originally isolated as an HIV-1 TAT-interactive protein, known to interact with the viral TAT protein that leads to HTATIP polyubiquitination and targets it for degradation. It belongs to the MYST family of histone acetyl transferases (HATs). TIP60 is a catalytic subunit of the NuA4 histone acetyltransferase complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histone H4 and H2A. A co-regulator of transcription factors, TIP60 either promotes or suppresses tumorigenesis. TIP60 is an enzyme that has a role in DNA repair and apoptosis and is thought to play an important role in signal transduction.
Specifications
| TIP60 | |
| Polyclonal | |
| Unconjugated | |
| KAT5 | |
| 60 kDa Tat-interactive protein; AI839539; cPLA(2)-interacting protein; CPLA2; cPLA2 interacting protein; ESA1; histone acetyltransferase HTATIP; Histone acetyltransferase KAT5; HIV-1 Tat interactive protein; HIV-1 tat interactive protein 1, 60 kDa homolog; HIV-1 tat interactive protein 1, homolog; HIV-1 Tat interactive protein 60 kD; HIV-1 Tat interactive protein, 60 kD; HIV-1 Tat interactive protein, 60kDa; HIV-1 tat interactive protein, homolog; HTATIP; Htatip1; K(lysine) acetyltransferase 5; K-acetyltransferase 5; Kat5; Lysine acetyltransferase 5; PLIP; Tat interacting protein, 60kDa; Tat-interactive protein-60; TIP; Tip55; Tip60; TIP60(HTATIP); Tip60b; ZC2HC5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10524, 192218, 81601 | |
| 4°C, do not freeze | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8CHK4, Q92993, Q99MK2 | |
| KAT5 | |
| A 15 amino acid peptide near the carboxy terminus of human KAT5. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction