missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIP47 Polyclonal Antibody

TIP47 Polyclonal Antibody
Brand: Invitrogen PA587949
This item is not returnable.
View return policy
Description
TIP47 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| TIP47 | |
| Polyclonal | |
| Unconjugated | |
| PLIN3 | |
| 1300012C15Rik; 47 kDa mannose 6-phosphate receptor-binding protein; 47 kDa MPR-binding protein; 47 kDa tail interaction protein; cargo selection protein TIP47; M6prbp1; mannose 6 phosphate receptor binding protein 1; mannose-6-phosphate receptor binding protein 1; mannose-6-phosphate receptor-binding protein 1; MGC11117; MGC2012; perilipin 3; perilipin-3; Placental protein 17; Plin3; PP17; tail-interacting protein, 47 kD; testicular tissue protein Li 114; TIP47; Unknown (protein for MGC:128897) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10226 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.16 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| O60664 | |
| PLIN3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 135-434 of human PLIN3 (NP_0058083). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction