missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIP30 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98510MFV450
This item is not returnable.
View return policy
Description
TIP30 Polyclonal antibody specifically detects TIP30 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| TIP30 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 30 kDa HIV-1 TAT-interacting protein, CC3oxidoreductase HTATIP2, EC 1.1.1, EC 1.1.1.-, FLJ26963, HIV-1 Tat interactive protein 2, 30 kDa, HIV-1 Tat interactive protein 2, 30kDa, HIV-1 TAT-interactive protein 2, SDR44U1, Tat-interacting protein (30kD), TIP30short chain dehydrogenase/reductase family 44U, member 1 | |
| Produced in rabbits immunized with E. coli-derived Human TIP30 fragment. | |
| 0.1 mL | |
| Apoptosis | |
| 10553 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction