missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIP120B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22334
This item is not returnable.
View return policy
Description
TIP120B Polyclonal specifically detects TIP120B in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| TIP120B | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| cullin-associated and neddylation-dissociated 2 (putative), Cullin-associated and neddylation-dissociated protein 2, cullin-associated NEDD8-dissociated protein 2, epididymis tissue protein Li 169, KIAA0667TBP-interacting protein of 120 kDa B, p120 CAND2, TBP interacting protein, TBP-interacting protein 120B, TIP120BTp120b | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg | |
| NP_001155971.1 | |
| CAND2 | |
| The immunogen this antibody was made to, maps to a region between residue 1186 to 1236 of human Cullin-Sssociated and Neddylation-Dissociated 2 using the numbering given in entry NP_001155971.1 (GeneID 23066). | |
| 100 μL | |
| Cell Cycle and Replication | |
| 23066 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction