missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIP120B Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22334APC
This item is not returnable.
View return policy
Description
TIP120B Polyclonal antibody specifically detects TIP120B in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| TIP120B | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| cullin-associated and neddylation-dissociated 2 (putative), Cullin-associated and neddylation-dissociated protein 2, cullin-associated NEDD8-dissociated protein 2, epididymis tissue protein Li 169, KIAA0667TBP-interacting protein of 120 kDa B, p120 CAND2, TBP interacting protein, TBP-interacting protein 120B, TIP120BTp120b | |
| The immunogen this antibody was made to, maps to a region between residue 1186 to 1236 of human Cullin-Sssociated and Neddylation-Dissociated 2 using the numbering given in entry NP_001155971.1 (GeneID 23066). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 23066 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction