missing translation for 'onlineSavingsMsg'
Learn More

TIP120B Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18402628 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402628 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402628 Supplier Novus Biologicals Supplier No. NBP222334AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TIP120B Polyclonal antibody specifically detects TIP120B in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIP120B
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias cullin-associated and neddylation-dissociated 2 (putative), Cullin-associated and neddylation-dissociated protein 2, cullin-associated NEDD8-dissociated protein 2, epididymis tissue protein Li 169, KIAA0667TBP-interacting protein of 120 kDa B, p120 CAND2, TBP interacting protein, TBP-interacting protein 120B, TIP120BTp120b
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1186 to 1236 of human Cullin-Sssociated and Neddylation-Dissociated 2 using the numbering given in entry NP_001155971.1 (GeneID 23066).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 23066
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.