missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiotropium-d6 Bromide, TRC
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Specifications
Specifications
| Chemical Name or Material | Tiotropium-d6 Bromide |
| Molecular Formula | C19H16D6BrNO4S2 |
| Quantity | 10 mg |
| Synonym | (1α,2β,4β,5α,7β)-7-[(2-Hydroxy-2,2-di-2-thienylacetyl)oxy]-9,9-dimethyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane-d6 Bromide |
| InChI Formula | InChI=1S/C19H22NO4S2.BrH/c1-20(2)12-9-11(10-13(20)17-16(12)24-17)23-18(21)19(22,14-5-3-7-25-14)15-6-4-8-26-15;/h3-8,11-13,16-17,22H,9-10H2,1-2H3;1H/q+1;/p-1/t11?,12-,13+,16-,17+;/i1D3,2D3; |
| SMILES | O=C(C(C1=CC=CS1)(O)C2=CC=CS2)O[C@H]3C[C@H]4[C@H]5[C@H](O5)[C@@H](C3)[N+]4(C([2H])([2H])[2H])C([2H])([2H])[2H].[Br-] |
| Molecular Weight (g/mol) | 478.45 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction