missing translation for 'onlineSavingsMsg'
Learn More

Tiotropium Bromide, MedChemExpress

Product Code. 16463317
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16463317

Brand: Medchem Express HY17360

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tiotropium Bromide (BA679 BR) is a muscarinic acetylcholine receptor (mAChR) antagonist that blocks the binding of the acetylcholine ligand and subsequent opening of the ligand-gated ion channel.

Tiotropium Bromide (BA679 BR) is a muscarinic acetylcholine receptor (mAChR) antagonist that blocks the binding of the acetylcholine ligand and subsequent opening of the ligand-gated ion channel.

Specifications

Chemical Name or Material Tiotropium Bromide
Physical Form Powder
CAS 136310-93-5
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H22BrNO4S2
For Use With (Application) Neuroscience-Neuromodulation
Synonym BA679 BR
Solubility Information DMSO : ≥ 30 mg/mL (63.50 mM) ∣H2O : 25 mg/mL (52.92 mM; Need ultrasonic)
SMILES O=C(C(C1=CC=CS1)(O)C2=CC=CS2)O[C@H]3C[C@H]([N+]4(C)C)[C@@H]5O[C@@H]5[C@H]4C3.[Br-]
Molecular Weight (g/mol) 472.42
Formula Weight 472.42
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.