missing translation for 'onlineSavingsMsg'
Learn More

Tiopinac, MedChemExpress

Product Code. 16463119
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16463119 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16463119 Supplier Medchem Express Supplier No. HYU00063
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tiopinac (RS 40974), a dibenzthiepin, is an orally active and highly potent anti-inflammatory and anti-pyretic agent.

Tiopinac (RS 40974), a dibenzthiepin, is an orally active and highly potent anti-inflammatory and anti-pyretic agent.

Specifications

Chemical Name or Material Tiopinac
CAS 61220-69-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H12O3S
Quantity 1 mg
Synonym RS 40974
SMILES O=C(O)CC1=CC=C2C(SCC3=CC=CC=C3C2=O)=C1
Molecular Weight (g/mol) 284.33
Formula Weight 284.33
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.