missing translation for 'onlineSavingsMsg'
Learn More

Tioconazole, TRC

Product Code. 30456657
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456657 10 g 10g
30450805 100 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30456657 Supplier Toronto Research Chemicals Supplier No. TRCT44468010G
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65899-73-2
Molecular Formula C16 H13 Cl3 N2 O S
Molecular Weight (g/mol) 387.71
Synonym Tioconazole, 1-[(2RS)-2-[(2-Chlorothiophen-3-yl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole
IUPAC Name 1-[2-[(2-chlorothiophen-3-yl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole
SMILES Clc1ccc(C(Cn2ccnc2)OCc3ccsc3Cl)c(Cl)c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C16H13Cl3N2OS/c17-12-1-2-13(14(18)7-12)15(8-21-5-4-20-10-21)22-9-11-3-6-23-16(11)19/h1-7,10,15H,8-9H2
Formula Weight 385.9814
Chemical Name or Material Tioconazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.