missing translation for 'onlineSavingsMsg'
Learn More

Tioconazole, MedChemExpress

Product Code. 16458448
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458448 10 g 10g
16440878 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16458448 Supplier Medchem Express Supplier No. HYB0319
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tioconazole (UK-20349) is an antifungal imidazole derivative with broad spectrum activity. Tioconazole has inhibitory active aginst several dermatophytes and several yeasts with MIC50s <3.12 mg/L and <9 mg/L, respectively.

Tioconazole (UK-20349) is an antifungal imidazole derivative with broad spectrum activity. Tioconazole has inhibitory active aginst several dermatophytes and several yeasts with MIC50s <3.12 mg/L and <9 mg/L, respectively.

Specifications

Chemical Name or Material Tioconazole
Physical Form Powder
CAS 65899-73-2
Color White
Quantity 10 g
Purity Grade Notes Research
Molecular Formula C16H13Cl3N2OS
For Use With (Application) COVID-19-immunoregulation
Synonym UK-20349
Solubility Information DMSO : 250 mg/mL (644.81 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.26 mM; Need ultrasonic)
SMILES ClC1=CC=C(C(OCC2=C(Cl)SC=C2)CN3C=CN=C3)C(Cl)=C1
Molecular Weight (g/mol) 387.71
Formula Weight 387.71
Percent Purity 99.8%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.