missing translation for 'onlineSavingsMsg'
Learn More

Cytiva TiO2 Mag Sepharose™

Product Code. 11590884 Shop All Cytiva Products
Click to view available options
Quantity:
500 μL
4 x 500 μL
Unit Size:
500µL
Pack of 4
This item is not returnable. View return policy

Product Code. 11590884

Brand: Cytiva 28951377

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed for enrichment of phosphopeptides from tryptic digested protein samples. Cytiva TiO2 Mag Sepharose magnetic beads are based on titanium dioxide chromatography and are designed to simplify capture and enrichment of phosphopeptides.

TiO2 Mag Sepharose has high affinity for phosphopeptides and is an efficient method for enrichment of phosphopeptides from complex samples. The magnetic bead format improves ease-of-use in sample preparation and has excellent properties for small-scale experiments. The high density beads allows rapid capture by magnetic devices with high visibility to ensure reliable collection of all bound target peptides.

  • Magnetic beads for selective capture and enrichment of phosphopeptides
  • Easy to use - visible and dense magnetic beads
  • Simple protocol with elution conditions optimized for MS analysis
  • Parallel sample handling when used with MagRack 6 separation tool - process up to six samples in less than one hour

 

TRUSTED_SUSTAINABILITY

Specifications

Bead Type TiO2 Mag Sepharose
Matrix Paramagnetic spherical, highly cross linked agarose particles
Particle Size 37 to 100 μm
Quantity 4 x 500 μL
Storage Buffer 20% ethanol storage solution
Content And Storage 4°C to 30°C (Use at room temperature)
For Use With (Application) Used for selective capture and enrichment of Phosphopeptides, for biomarker research
Ligand TiO2
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.