missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ VARIO™ Sulfate 4 Reagent, 25mL
Click to view available options
Quantity:
25 mL
Unit Size:
Pack of 1000
Description
Formulated for use in Lovibond test kits, colorimeters and spectrophotometers.
- For use in our test kits, colorimeters and spectrophotometers
- Pre-measured, unit dose reagents
- Easy to handle, easy to open and easy to pour
- Case of 100
- Fast and easy use of VARIO Powder Packs has made them extremely popular for water testing applications in many countries throughout the world
- Provides more experienced users with a real alternative to existing measurement systems
- Produced to the same high quality standards that have made Tintometer’s tablet reagents so successful for several decades
- Parameters from aluminum and chlorine through to sulphate are just some of the well-known tests that are included in the VARIO Powder Pack range
Specifications
Specifications
| Format | Powdered Reagent |
| For Use With (Application) | Water Testing |
| For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
| Quantity | 25 mL |
| Type | VARIO™ Sulfate 4 Reagent |
Safety and Handling
Product Identifier
- X1000 POWDER VARIO SULFA 4 25ML .
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H301-Toxic if swallowed.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- MIXTURE LIST-Contains : citric acid, bariumchloride dihydrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction