missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ VARIO™ Silica Ultra HR Reagent Set, 25mL

Product Code. 15391695
Click to view available options
Quantity:
25 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15391695

Brand: Lovibond™ 535900

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Formulated for use in Lovibond test kits, colorimeters and spectrophotometers.

  • Range: 0 to 200mg/L
  • Parameter: Silica
  • For use in our test kits, colorimeters and spectrophotometers
  • Case of 100

Specifications

For Use With (Application) Water Testing
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 25 mL
Type Silica Ultra HR Reagent Set
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.