missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ VARIO™ Molybdenum HR1/HR2/HR3 Powder Reagent Set
Click to view available options
Quantity:
100 Tests
Unit Size:
10mL
Description
- For use in our test kits, colorimeters and spectrophotometers
- Pre-measured, unit dose reagents
- Easy to handle, easy to open and easy to pour
- Case of 100
- Fast and easy use of VARIO Powder Packs has made them extremely popular for water testing applications in many countries throughout the world
- Provides more experienced users with a real alternative to existing measurement systems
- Produced to the same high quality standards that have made Tintometer’s tablet reagents so successful for several decades
- Parameters from aluminum and chlorine through to sulphate are just some of the well-known tests that are included in the VARIO Powder Pack range
Specifications
Specifications
| For Use With (Application) | Water Testing |
| For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
| Quantity | 100 Tests |
| Type | VARIO™ Molybdenum HR1/HR2/HR3 Powder Reagent Set |
Safety and Handling
Product Identifier
- 10ML VARIO MOLYBDENUM HR SET .
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : succinic acid, glycene, disodium peroxodisulphate, calcium thioglycolate trihydrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction