Learn More
Tintometer™ Vario Manganese Reagent Set
For use with Lovibond™ Tintometer photometers
Brand: Tintometer™ 535090
Code : NEW
Additional Details : Weight : 0.12000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Description
- Parameter: Manganese Reagent LR F10
- Includes: Vario alkaline-cyanide solution (60mL), Vario ascorbic acid powder pack (100/Pk.), Vario PAN indicator solution (60mL)
Specifications
Manganese Reagent LR F10 | |
Vario alkaline-cyanide solution (60mL), Vario ascorbic acid powder pack (100/Pk.), Vario PAN indicator solution (60mL) | |
Reagent Set |
Safety and Handling
- vario manganese reagent set LR
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H226-Flammable liquid and vapour.
- H290-May be corrosive to metals.
- H301-Toxic if swallowed.
- H310-Fatal in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H360D-May damage the unborn child.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H411-Toxic to aquatic life with long lasting effects.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : sodium cyanide, sodium hydroxide, N,N-dimethylformamide, octylphenol polyrhtoxyethanol