missing translation for 'onlineSavingsMsg'
Learn More

Tintometer™ Vario Manganese Reagent Set

Product Code. 11698318
Click to view available options
:
Manganese Reagent LR F10
This item is not returnable. View return policy

Product Code. 11698318

Brand: Tintometer™ 535090

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use with Lovibond™ Tintometer photometers

  • Parameter: Manganese Reagent LR F10
  • Includes: Vario alkaline-cyanide solution (60mL), Vario ascorbic acid powder pack (100/Pk.), Vario PAN indicator solution (60mL)

Specifications

Format Manganese Reagent LR F10
Includes Vario alkaline-cyanide solution (60mL), Vario ascorbic acid powder pack (100/Pk.), Vario PAN indicator solution (60mL)
Type Reagent Set
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • vario manganese reagent set LR
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 3
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Reproductive toxicity Category 1B
  • Substances and mixtures corrosive to metals Category 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H360D-May damage the unborn child.
  • H310-Fatal in contact with skin.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H400-Very toxic to aquatic life.
  • H290-May be corrosive to metals.
  • H411-Toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : sodium cyanide, sodium hydroxide, N,N-dimethylformamide, octylphenol polyrhtoxyethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.