missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ VARIO™ Iron Ferro Reagent Powder, 25mL
Click to view available options
Quantity:
25 mL
Unit Size:
Pack of 1000
Description
- For use in our test kits, colorimeters and spectrophotometers
- Pre-measured, unit dose reagents
- Easy to handle, easy to open and easy to pour
- Case of 100
- Fast and easy use of VARIO Powder Packs has made them extremely popular for water testing applications in many countries throughout the world
- Provides more experienced users with a real alternative to existing measurement systems
- Produced to the same high quality standards that have made Tintometer’s tablet reagents so successful for several decades
- Parameters from aluminum and chlorine through to sulphate are just some of the well-known tests that are included in the VARIO Powder Pack range
Specifications
Specifications
| For Use With (Application) | Water Testing |
| For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
| No. of Tests | Case of 1000 |
| Quantity | 25 mL |
| Type | VARIO™ Iron Ferro Reagent Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction