missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ Urea Reagent 2, 10mL
Click to view available options
Quantity:
10 mL
Unit Size:
15mL
Description
- Urea Reagent 2, 10mL
- Photometry Liquid Reagent
- As a rule, liquid reagents do not consist of a single preparation but comprise several components that need to be added to the sample in a certain order. As both the size and the number of drops have a decisive effect on the resultant color complex, the reagents need to be added with a high degree of precision. The useful life of liquid reagents is reduced by temporary contact with oxygen in the air when the bottle is opened as well as by unsuitable storage environments (presence of sunlight or high temperatures).
Specifications
Specifications
| For Use With (Application) | Photometry Liquid Reagent |
| For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
| Quantity | 10 mL |
| Type | Urea Reagent 2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction