missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ Tannin No. 2 Tablets, 2 to 20mg/L

Product Code. 15361995
Click to view available options
Quantity:
100 Tablets
Unit Size:
Pack of 250
This item is not returnable. View return policy

Product Code. 15361995

Brand: Lovibond™ 503511

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits

  • Range: 2 to 20mg/L or tannin units
  • Parameter: Organophosphonate
  • Method: Tablet count
  • Tablet reagents have a minimum 5 year shelf life
  • Case of 100

Specifications

For Use With (Application) Photometry Tablet Reagents
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 100 Tablets
Range 2 to 20mg/L or tannin units
Test Method Tablet count
Type Tannin No. 2 Tablets
compliance-icons compliance-icons
Product Identifier
  • X250 TANNIN NO.2 TABLETS
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : potassium permanganate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.