Learn More
Lovibond™ Tintometer™ Tannin No. 1 Tablets, 2 to 20mg/L
Click to view available options
Quantity:
100 Tablets
Unit Size:
Pack of 100
Description
- Range: 2 to 20mg/L or tannin units
- Parameter: Organophosphonate
- Method: Tablet count
- Tablet reagents have a minimum 5 year shelf life
- Case of 100
Specifications
Specifications
| For Use With (Application) | Photometry Tablet Reagents |
| For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
| Quantity | 100 Tablets |
| Range | 2 to 20mg/L or tannin units |
| Test Method | Tablet count |
| Type | Tannin No. 1 Tablets |