Learn More
Lovibond™ Tintometer™ Standard Solution Phosphate, 4.6mg/L PO4 = 1.5mg/L P
Click to view available options
:
Standard Solution Ammonia
Description
- Standard Solution Phosphate
- 4.6mg/L PO4 = 1.5mg/L P
- For use with Lovibond test kits, colorimeters and spectrophotometers
Specifications
Specifications
| Description | 4.6mg/L PO4 = 1.5mg/L P |
| For Use With (Equipment) | Test Kits, Colorimeters and Spectrophotometers |
| Type | Phosphate Solution |