Learn More
Lovibond™ Tintometer™ Standard Solution Phosphate, 20mg/l PO4 = 6.5mg/l P
For use with Lovibond test kits, colorimeters and spectrophotometers
Brand: Lovibond™ 2420809
Code : NEW
Additional Details : Weight : 0.30000kg
Description
- Standard Solution Phosphate
- 20mg/l PO4 = 6.5mg/l P
- For use with Lovibond test kits, colorimeters and spectrophotometers
Specifications
20mg/l PO4 = 6.5mg/l P | |
Test Kits, Colorimeters and Spectrophotometers | |
Phosphate Solution |