missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ SensoDirect 150 All-In-One Handheld Meter, pH/Con/TDS

Product Code. 12611745
Click to view available options
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12611745

Brand: Lovibond™ 724210

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Features include automatic calibration, buffer recognition, large LCD display and result hold function. Lovibond SensoDirect 150 is combined instrument capable of measuring pH, ORP, Conductivity, TDS, Dissolved Oxygen and Temperature in a single meter.

  • Test: pH Value; Range: 0 to 14
  • Test: Conductivity; Range: 0 to 200uS/cm; 0.2 to 200mS/cm
  • SensoDirect 150 meter
  • Battery, 9V
  • pH electrode, plastic, type 110
  • Conductivity Electrode
  • pH buffer set with pH 4.0 and pH 7
  • Conductivity standard 1413 uS/cm
  • Instruction manual and guarentee sheet

Specifications

Accuracy (pH) 0 to 14 pH, ±0.02 pH + 2 digits
Accuracy (mV) 0 to 1999mV, 1mV
Accuracy (Temperature) ±0.8°C (1.5°F)
Type SensoDirect 150 Set pH/Con/TDS
Dimensions (L x W x H) 220 x 120 x 40 mm
For Use With (Application) Measures pH/Redox, dissolved oxygen and conductivity/TDS
pH Range 0 to 14
Range (Conductivity) 0 to 200.0 µS
Range (mV) mV to 1999mV to 1999mV
Range (TDS) 0 to 132,000ppm
Resolution (Conductivity) (Range/Resolution) 0 to 200.0µS / 0.1µS; 0.2 to 2.000mS / 0.001mS; 2 to 20.00mS / 0.01mS; 20 to 200.00mS / 0.1mS
Resolution (mV) 0 to 1999mV, 1mV
Resolution (pH) 0 to 14pH, 0.01 pH
Resolution (Temperature) 0.1°C
Temperature Compensation Manual: 0°C to 100°C, Automatic (ATC)
Temperature (Metric) Range 0°C to 50°C
Weight (Metric) 625 g
Certifications/Compliance CE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.