missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ SD 320 Con Waterproof Handheld Meter

Product Code. 15410260
Change view
Click to view available options
Warranty:
Yes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Warranty unitSize
15410260 Yes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15410260 Supplier Lovibond™ Supplier No. 724700

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Measures Conductivity, Salinity, TDS and Temperature. The SD 300 series is a range of waterproof hand-held meters for drinking water, cooling and boiler water, waste water, pool water and surface water applications.

  • Contains the instrument and Conductivity Cell LC 12 (measuring range 0 to 200 mS/cm), batteries, manual and warranty information
  • Min / Max Value Memory: highest and lowest measured value is saved
  • Auto Hold freeze and display measurement
  • Auto Power Off: if unused, the meter automatically switches off after a selected period (0 to 120 min., or deactivated)
  • Automatic temperature compensation
  • Salinity measurement
  • TDS measurement (total dissolved solids)
  • Rugged, water resistant (IP 67) designed for field use
  • PC interface (USB / serial or analog)
  • Automatic buffer detection (SD 300 pH)
  • Data logger and alarm function (min./max.)
  • Good Laboratory Practice (GLP-features)
  • Clear, concise result reading on easy-to-read backlit LCD display
  • Automatic temperature compensation
  • High resolution
  • Dirt-insensitive up-to-date 4-pole conductivity cell offering highest precision (SD 320 Con)

Specifications

Includes Instrument, batteries, conductivity cell LC 12, manual
Accuracy (Temperature) ±0.2°C
Temperature Range (Metric) -5.0°C to +150°C
Display Type Two, 4.5 Digit Seven-Segment Displays (15 mm and 12 mm)
IP Rating IP67 (housing and connections)
Dimensions (L x W x H) 37 x 164 x 128 mm
Warranty Yes
Accuracy (Conductivity) ±0.5% of reading; ±0.1% FS (depends on electrode)
Model SD 320 Con
Power Supply 2 x AAA-battery (included); Power consumption: < 6,25mA
Range (Resistivity) 0.005 to 500.0kOhm/cm
Range (Salinity) 0.0 to +70.0 (g salt / kg water
Range (TDS) 0 to +5000mg/L
Type SD 320 Con Waterproof Hand-held Meter
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.