missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ Potassium Iodide Tablets

Product Code. 15311985
Change view
Click to view available options
Quantity:
100 Tablets
250 Tablets
Unit Size:
Pack of 100
Pack of 250
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15311985 100 Tablets Pack of 100
15321985 250 Tablets Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15311985 Supplier Lovibond™ Supplier No. 501210
  • ONLINE EXCLUSIVE PRICE
    £20.10
    £17.88 / Pack of 100
    Save £2.22
    11% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits

  • Potassium Iodide Tablets
  • Available in bottles of 100 and 250

Specifications

For Use With (Application) Water Testing
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 100 Tablets
Type Potassium Iodide Tablets
compliance-icons
Product Identifier
  • X100 POTASSIUM IODIDE TABLETS .
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P314-Get medical advice/attention if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: potassium iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.