Learn More
Lovibond™ Tintometer™ Phosphate (Ortho) 2 Reagent Ampoule, 0.05 to 5mg/L
Click to view available options
No. of Tests:
30
Unit Size:
Each
Description
- Range: 0.05 to 5mg/L
- Parameter: Ortho Phosphate
- Method: Stannous Chloride
- For use in test kits, colorimeters and spectrophotometers
- Case of 30
Specifications
Specifications
| For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
| No. of Tests | 30 |
| Range | 0.05 to 5mg/L |
| Test Method | Stannous Chloride |
| Type | Phosphate (Ortho) 2 Reagent Ampoule |