missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ Phenol Red Solution, 15mL

Product Code. 15352015
Click to view available options
Quantity:
15 mL
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 15352015

Brand: Lovibond™ 471046

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use with Lovibond test kits, colorimeters and spectrophotometers

  • Phenol Red Solution
  • 15mL
  • Pack of 6 Bottles

Specifications

For Use With (Application) pH Test
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 15 mL
Type Phenol Red Solution
compliance-icons
Product Identifier
  • SOLUTION ROUGE DE PHENOL 15 ML
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P233-Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : butan-2-ol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.