missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ Phenol No. 2 Tablet

Product Code. 10364493
Click to view available options
Quantity:
100 Tablets
Unit Size:
100 tablets
This item is not returnable. View return policy

Product Code. 10364493

Brand: Lovibond™ 515960BT

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits

  • New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
  • With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
  • Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
  • Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
  • ‘BT’ is added to the end of the code to identify the new style of packaging
  • Simple, time-saving and practical
  • There are no safety risks if the tablets are used in line with the instructions supplied
  • Safety data sheets are available for all reagents

Specifications

For Use With (Application) Photometry Tablet Reagents
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 100 Tablets
Type Phenol No. 2 Tablet
compliance-icons compliance-icons
Product Identifier
  • phenole no. 2
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : ammonium persulphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.