missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ Molybdate No. 1 HR Tablet
Click to view available options
Quantity:
100 Tablets
250 Tablets
Unit Size:
Pack of 100
Pack of 250
Description
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
- Range: 1 to 50mg/L
- Parameter: Molybdate LR, molybdate, HR molybdate
- Method: Thioglycolate
- For use in our test kits, colorimeters and spectrophotometers
- New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
- With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
- Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
- Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
- ‘BT’ is added to the end of the code to identify the new style of packaging
- Simple, time-saving and practical
- There are no safety risks if the tablets are used in line with the instructions supplied
- Safety data sheets are available for all reagents
Specifications
Specifications
| For Use With (Application) | Photometry Tablet Reagents |
| For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
| Quantity | 250 Tablets |
| Range | 1 to 50mg/L |
| Type | Molybdate No. 1 HR Tablet |
Safety and Handling
Product Identifier
- X250 MOLYBDATE NO.1 TABLETS .
- Danger
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : adipic acid, sodiumbisulphate, potassium persulphate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction