Learn More
Lovibond™ Tintometer™ Hydrazine Vacu-vial™ Reagent Set, 0.01 to 0.7mg/L
Click to view available options
No. of Tests:
30
Unit Size:
Each
Description
- Range: 0.01 to 0.7mg/L
- Parameter: Hydrazine
- Method: PDMAB
- For use in test kits, colorimeters and spectrophotometers
- Case of 30
Specifications
Specifications
| For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
| No. of Tests | 30 |
| Range | 0.01 to 0.7mg/L |
| Test Method | PDMAB |
| Type | Hydrazine Vacu-vial™ Reagent Set |