missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ DPD No 4 Tablets
Click to view available options
Quantity:
25 Tablets
500 Tablets
Unit Size:
Pack of 250
Pack of 500
Description
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
- DPD No. 4 Tablets
- Available in packs of 250 and 500
- New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
- With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
- Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
- Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
- ‘BT’ is added to the end of the code to identify the new style of packaging
- Simple, time-saving and practical
- There are no safety risks if the tablets are used in line with the instructions supplied
- Safety data sheets are available for all reagents
Specifications
Specifications
| For Use With (Application) | Photometry Tablet Reagents |
| For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
| Quantity | 500 Tablets |
| Type | DPD No 4 Tablets |
Safety and Handling
Signal Word
- Danger
- Specific target organ toxicity Category 1
- H372-Causes damage to organs.
- P264-Wash thoroughly after handling.
- P314-Get medical advice/attention if you feel unwell.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction