Learn More
Lovibond™ Tintometer™ DPD No. 3 Tablets
Product Code. 15361935
Change view
Click to view available options
Quantity:
250 Tablets
Unit Size:
Pack of 250
Product Code. Quantity Unit Size
15361935 250 Tablets Pack of 250
1 options
Product Code. 15361935 Supplier Lovibond™ Supplier No. 511081BT
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
  • DPD No. 3 Tablets
  • 250 per pack
  • New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
  • With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
  • Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
  • Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
  • ‘BT’ is added to the end of the code to identify the new style of packaging
  • Simple, time-saving and practical
  • There are no safety risks if the tablets are used in line with the instructions supplied
  • Safety data sheets are available for all reagents
Specifications
For Use With (Application) Photometry Tablet Reagents
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 250 Tablets
Type DPD No. 3 Tablets