Learn More
Lovibond™ Tintometer™ DPD No. 3 High Calcium Tablets
Click to view available options
Quantity:
100 Tablets
250 Tablets
500 Tablets
Unit Size:
Pack of 100
Pack of 250
Pack of 500
Description
- New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
- With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
- Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
- Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
- ‘BT’ is added to the end of the code to identify the new style of packaging
- Simple, time-saving and practical
- There are no safety risks if the tablets are used in line with the instructions supplied
- Safety data sheets are available for all reagents
Specifications
Specifications
| For Use With (Application) | Photometry Tablet Reagents |
| For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
| Quantity | 250 Tablets |
| Type | DPD No. 3 High Calcium Tablets |
Safety and Handling
Product Identifier
- X250 DPD NO.3 HIGH CALCIUM TABLETS .
- Danger
- Specific target organ toxicity Category 1
- H372-Causes damage to organs.
- P264-Wash hands thoroughly after handling.
- P314-Get medical advice/attention if you feel unwell.
- MIXTURE LIST-Contains : potassium iodide