Learn More
Lovibond™ Tintometer™ DPD Nitrite Tablets
Product Code. 15341995
Click to view available options
Quantity:
250 Tablets
Unit Size:
Pack of 250
Product Code. 15341995
Brand: Lovibond™ 502691
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
  • DPD Nitrite Tablets
  • Free and combined
  • Bottle of 250
Specifications
Analytes DPD Nitrite Tablets (free and combined)
For Use With (Application) Water Testing
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 250 Tablets
Type Photometry Reagent