Learn More
Lovibond™ Tintometer™ DPD Acidifying Comparator Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand: Lovibond™ 502111
Code : NEW
Additional Details : Weight : 0.30000kg
Description
- DPD Acidifying Comparator Tablets
- Bottle of 250
- Test tablets are manufactured in Germany under tightly controlled conditions on most modern machinery. Maintaining the highest quality standards permits Tintometer to guarantee our tablet reagents for a minimum of 5 years, and some for as long as 10 years.
Specifications
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
250 Tablets | |
DPD Acidifying Comparator Tablets |