missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ DPD 3 Reagent Solution, 100mL
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
For use with Lovibond test kits, colorimeters and spectrophotometers
- DPD 3 Solution
- Black Bottle
- Red Cap
- 100mL
Specifications
Specifications
| For Use With (Application) | Water Testing |
| For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
| Quantity | 100 mL |
| Type | DPD 3 Reagent Solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction