missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ Dissolved Oxygen Reagent No 1, 30mL

Product Code. 15362035
Click to view available options
Quantity:
30 mL
Unit Size:
30mL
This item is not returnable. View return policy

Product Code. 15362035

Brand: Lovibond™ 461150

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use with Lovibond test kits, colorimeters and spectrophotometers

  • Dissolved Oxygen Reagent No 1, 30mL
As a rule, liquid reagents do not consist of a single preparation but comprise several components that need to be added to the sample in a certain order. As both the size and the number of drops have a decisive effect on the resultant color complex, the reagents need to be added with a high degree of precision. The useful life of liquid reagents is reduced by temporary contact with oxygen in the air when the bottle is opened as well as by unsuitable storage environments (presence of sunlight or high temperatures).

Specifications

For Use With (Application) Dissolved Oxygen Test
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
No. of Tests 100
Quantity 30 mL
Type Dissolved Oxygen Reagent No 1
compliance-icons
Product Identifier
  • 30ML FOR 100 TESTS OXYGEN REAGENT 1 .
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : manganese sulphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.