Learn More
Lovibond™ Tintometer™ Comparator 2000+ Disc 3/117, Iron Test, 1 to 10ppm
Description
- The required accuracy of results is only ensured if stable, fade-free color standards are used
- Glass color standards are fade-free, resistant to chemicals and scratchproof
- Lovibond™ standards are made from colored glass filters
- Comply with international standards, e.g. ISO 7393/2
- Readings: 1, 2, 3, 4, 5, 6, 7, 8, 10mg/L
Specifications
Specifications
| Analytes | Iron Test |
| For Use With (Equipment) | Comparator 2000+ |
| Range | 1 to 10ppm |
| Type | Comparator 2000+ Disc 3/117 |