missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ Chlorine HR Tablets (Potassium Iodide 0. 1g)
Click to view available options
Quantity:
250 Tablets
Unit Size:
Pack of 250
Description
- Chlorine HR Tablets (Potassium Iodide, 0. 1g)
- New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
- With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
- Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
- Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
- ‘BT’ is added to the end of the code to identify the new style of packaging
- Simple, time-saving and practical
- There are no safety risks if the tablets are used in line with the instructions supplied
- Safety data sheets are available for all reagents
Specifications
Specifications
| For Use With (Application) | Photometry Tablet Reagents |
| For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
| Quantity | 250 Tablets |
| Type | Chlorine HR Tablets (Potassium Iodide 0. 1g) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction