missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ Chlordioxide No.2 Tablets

Product Code. 15341985
Click to view available options
Quantity:
250 Tablets
Unit Size:
Pack of 250
This item is not returnable. View return policy

Product Code. 15341985

Brand: Lovibond™ 501281

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits

  • Chlordioxide No.2 Tablets
  • Bottle of 250
Test tablets are manufactured in Germany under tightly controlled conditions on most modern machinery. Maintaining the highest quality standards permits Tintometer to guarantee our tablet reagents for a minimum of 5 years, and some for as long as 10 years.

Specifications

For Use With (Application) Photometry Tablet Reagents
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 250 Tablets
Type Chlordioxide No.2 Tablets
compliance-icons
Product Identifier
  • X250 CHLORDIOXIDE NO.2 TABLETS
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P314-Get medical advice/attention if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : potassium iodide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.