Learn More
Lovibond™ Tintometer™ Calcium Hardness Tablets
Product Code. 15302005
Click to view available options
:
Calcium Hardness Tablets
Product Code. 15302005
Brand: Lovibond™ 505191
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
  • Calcium Hardness Tablets
  • Method: Tablet Count
  • Bottle of 250
Specifications
For Use With (Application) Calcium Hardness Test
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Test Method Tablet Count
Type Calcium Hardness Tablets