Learn More
Lovibond™ Tintometer™ Alkalinity Total M Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand: Lovibond™ 505321
Code : NEW
Additional Details : Weight : 0.30000kg
Beschreibung
- Alkalinity Total M Tablets
- Method: Tablet Count
- Bottle of 250
Spezifikation
Photometry Tablet Reagents | |
250 Tablets | |
Alkalinity Total M Tablets |
Test kits, colorimeters and spectrophotometers | |
Tablet Count |