missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ Active Oxygen, Copper and pH Refill Reagent Packs
Click to view available options
:
Active Oxygen, Copper and pH Refill Reagent Packs
Description
- Refill Reagent Packs: Active Oxygen, Copper and pH
- Contains: 20 (DPD) No. 4 Rapid Tablets, 20 Copper No. 1 Tablets And 30 Phenol Red Rapid Tablets
Specifications
Specifications
| For Use With (Application) | Photometry Tablet Reagents |
| For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
| Includes | 20 (DPD) No. 4 Rapid Tablets, 20 Copper No. 1 Tablets And 30 Phenol Red Rapid Tablets |
| Test Method | Azomethine |
| Type | Active Oxygen, Copper and pH Refill Reagent Packs |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction