Learn More
Lovibond™ Tintometer™ Acidifying PT Rapid Tablets
Product Code. 15311855
Click to view available options
Quantity:
100 Tablets
250 Tablets
Unit Size:
Pack of 100
Pack of 250
Product Code. 15311855
Brand: Lovibond™ 515490BT
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
  • Acidifying PT Rapid Tablets
  • Available in packs of 100 and 250
Specifications
For Use With (Application) Photometry Tablet Reagents
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 100 Tablets
Type Acidifying PT Rapid Tablets