missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TINP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16230-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the ubiquitin-specific processing (UBP) family of proteases that is a deubiquitinating enzyme (DUB) with His and Cys domains. This protein is located in the cytoplasm and cleaves the ubiquitin moiety from ubiquitin-fused precursors and ubiquitinylated proteins. Mice with a mutation that results in reduced expression of the ortholog of this protein are retarded for growth, develop severe tremors by 2 to 3 weeks of age followed by hindlimb paralysis and death by 6 to 10 weeks of age. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
| TINP1 | |
| Polyclonal | |
| Unconjugated | |
| NSA2 | |
| CDK105, Hairy cell leukemia protein 1, HCL G1, HUSSY29, NSA2, TINP1, YR 29 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10412, 171456, 59050 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O95478, Q9CR47, Q9QYU7 | |
| Nsa2 | |
| TINP1 Fusion Protein Ag8822 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction