missing translation for 'onlineSavingsMsg'
Learn More

TINP1 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18424844 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424844 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424844 Supplier Novus Biologicals Supplier No. NBP177374AF350

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chicken Polyclonal Antibody

TINP1 Polyclonal antibody specifically detects TINP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TINP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias FLJ94393, Hairy cell leukemia protein 1, HCLG1, HCL-G1, HUSSY29, HUSSY-29, NSA2 ribosome biogenesis homolog (S. cerevisiae), ribosome biogenesis protein NSA2 homolog, TGF beta-inducible nuclear protein 1, TGF-beta inducible nuclear protein, TGF-beta-inducible nuclear protein 1, TINP1CDK105, YR-29
Host Species Chicken
Immunogen Antibody was raised against a 13 amino acid synthetic peptide near the carboxy terminus of human TINP1. The immunogen is located within the last 50 amino acids of TINP1. Amino Acid Squence: KYAQVTNNPENDG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10412
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.