missing translation for 'onlineSavingsMsg'
Learn More

Tinidazole, MedChemExpress

Product Code. 16418156
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16418156 100 mg 100mg
16412079 1 g 1g
16400808 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16418156 Supplier Medchem Express Supplier No. HYB0177
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tinidazole, an orally available antibacterial agent, is a 5-nitroimidazole with selective activity against anaerobic bacteria and protozoa.

Tinidazole, an orally available antibacterial agent, is a 5-nitroimidazole with selective activity against anaerobic bacteria and protozoa.

Specifications

Chemical Name or Material Tinidazole
Physical Form Solid
CAS 19387-91-8
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H13N3O4S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 50 mg/mL (202.21 mM) ∣H2O : 3.33 mg/mL (13.47 mM; Need ultrasonic)
SMILES CC1=NC=C([N+]([O-])=O)N1CCS(=O)(CC)=O
Molecular Weight (g/mol) 247.27
Formula Weight 247.27
Percent Purity 99.87%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.