missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TINAGL1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA561805

Product Code. 15759611

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: DQEPCLVDPD MIKAINQGNY GWQAGNHSAF WGMTLDEGIR YRLGTIRPSS SVMNMHEIYT VLNPGEVLPT AFEASEKWPN LI Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

TINAGL1
Polyclonal
Unconjugated
TINAGL1
1110021J17Rik; Adrenocortical zonation factor 1; androgen-regulated gene 1; androgen-regulated gene 1 protein; Arg1; AZ1; AZ-1; Gis5; glucocorticoid-inducible protein 5; LCN7; LIECG3; lipocalin 7; OLRG2; OLRG-2; Oxidized LDL-responsive gene 2 protein; oxidized-LDL responsive gene 2; P3ECSL; PP6614; PSEC0088; TARP; TIN Ag-related protein; Tinagl; TINAGL1; TINAG-like 1; TINAGRP; TIN-Ag-RP; tubulointerstitial nephritis antigen like 1; tubulointerstitial nephritis antigen-like; tubulointerstitial nephritis antigen-like 1; tubulointerstitial nephritis antigen-related protein; UNQ204/PRO230
Rabbit
Antigen affinity chromatography
RUO
64129
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9GZM7
TINAGL1
Recombinant protein corresponding to Human TINAGL1. Recombinant protein control fragment (Product #RP-100961).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.