missing translation for 'onlineSavingsMsg'
Learn More

TINAGL1 Antibody [Biotin], Novus Biologicals™

Product Code. 30733074 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733074 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733074 Supplier Novus Biologicals Supplier No. AF7185B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

TINAGL1 Polyclonal antibody specifically detects TINAGL1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TINAGL1
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias androgen-regulated gene 1, ARG1, GIS5, Glucocorticoid-inducible protein 5, LCN7, LIECG3, lipocalin 7, OLRG2, OLRG-2, Oxidized LDL-responsive gene 2 protein, oxidized-LDL responsive gene 2, P3ECSL, TIN Ag-related protein, TINAGL, TINAG-like 1, TIN-Ag-RP, TINAGRP, tubulointerstitial nephritis antigen-like, tubulointerstitial nephritis antigen-like 1, Tubulointerstitial nephritis antigen-related protein
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human TINAGL1, Ala22-Met464, Accession # Q9GZM7
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 64129
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.